Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178907.3 | NP_849238.1 | 9 | 1271 | ||
GenBank mRNA | AK087496.1 | - | 8 | 932 | ||
AK144637.1 | - | 9 | 1209 | |||
AK151881.1 | - | 9 | 1194 | |||
AK170456.1 | - | 9 | 1267 | |||
AK172344.1 | - | 9 | 1182 | |||
BC031467.1 | - | 9 | 1271 | 4189474 |