Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178907.3 | NP_849238.1 | 8 | 1214 | ||
GenBank mRNA | AK051962.1 | - | 7 | 2222 | ||
AK087496.1 | - | 7 | 875 | |||
AK144637.1 | - | 8 | 1152 | |||
AK151881.1 | - | 8 | 1137 | |||
AK170456.1 | - | 8 | 1210 | |||
AK172344.1 | - | 8 | 1125 | |||
BC031467.1 | - | 8 | 1214 | 4189474 |