Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006187.3 | NP_006178.2 | 8, 9 | 1939 | ||
XM_005253889.3 | XP_005253946.1 | 8, 9 | 1860 | |||
XM_017019363.1 | XP_016874852.1 | 8, 9 | 1860 | |||
GenBank mRNA | AB044545.1 | - | 8, 9 | 2020 | ||
AF063613.1 | - | 8, 9 | 1875 | |||
AK292949.1 | - | 8, 9 | 1909 | |||
BC113746.1 | - | 8, 9 | 1914 | 40036464 | ||
BM474470.1 | - | 3, 4 | 637 |