Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153537.4 | NP_705765.3 | 20 | 3967 | ||
GenBank mRNA | AK052352.1 | - | 9 | 1327 | ||
AK122336.1 | - | 18 | 3886 | |||
BC025856.1 | - | 2 | 165 | |||
BC053931.1 | - | 15 | 2096 | |||
BC058712.1 | - | 20 | 3963 | 6842596 | ||
BC069853.1 | - | 1 | 94 |