Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000274.3 | NP_000265.1 | 3 | 454 | ||
NM_001171814.1 | NP_001165285.1 | 2 | 226 | |||
NM_001322965.1 | NP_001309894.1 | 3 | 449 | |||
NM_001322966.1 | NP_001309895.1 | 4 | 769 | |||
NM_001322967.1 | NP_001309896.1 | 4 | 574 | |||
NM_001322968.1 | NP_001309897.1 | 5 | 661 | |||
NM_001322969.1 | NP_001309898.1 | 4 | 541 | |||
NM_001322970.1 | NP_001309899.1 | 5 | 757 | |||
NM_001322974.1 | NP_001309903.1 | 2 | 226 | |||
XM_017016279.1 | XP_016871768.1 | 3 | 396 | |||
GenBank mRNA | AB208817.1 | - | 3 | 385 | ||
AK296032.1 | - | 2 | 173 | |||
AK312561.1 | - | 3 | 401 | |||
AK315947.1 | - | 2 | 173 | |||
BC000964.2 | - | 3 | 382 | 3449883 | ||
BC016928.1 | - | 3 | 386 | 3913352 | ||
CD104356.1 | - | 3 | 397 | |||
CR457045.1 | - | 2 | 321 | |||
CR749808.1 | - | 2 | 176 | |||
M12267.1 | - | 3 | 375 | |||
M14963.1 | - | 2 | 365 | |||
M23204.1 | - | 3 | 372 | |||
Y07511.1 | - | 2 | 361 |