Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000274.3 | NP_000265.1 | 9 | 1227 | ||
NM_001171814.1 | NP_001165285.1 | 8 | 999 | |||
NM_001322965.1 | NP_001309894.1 | 9 | 1222 | |||
NM_001322966.1 | NP_001309895.1 | 10 | 1542 | |||
NM_001322967.1 | NP_001309896.1 | 10 | 1347 | |||
NM_001322968.1 | NP_001309897.1 | 11 | 1434 | |||
NM_001322969.1 | NP_001309898.1 | 10 | 1314 | |||
NM_001322970.1 | NP_001309899.1 | 11 | 1530 | |||
NM_001322971.1 | NP_001309900.1 | 7 | 906 | |||
NM_001322974.1 | NP_001309903.1 | 9 | 1113 | |||
XM_017016279.1 | XP_016871768.1 | 9 | 3022 | |||
GenBank mRNA | AB208817.1 | - | 9 | 3012 | ||
AK296032.1 | - | 8 | 946 | |||
AK312561.1 | - | 9 | 1174 | |||
AK315947.1 | - | 8 | 946 | |||
BC000964.2 | - | 9 | 1155 | 3449883 | ||
BC016928.1 | - | 9 | 1159 | 3913352 | ||
CR457045.1 | - | 8 | 1094 | |||
CR749808.1 | - | 8 | 949 | |||
M12267.1 | - | 9 | 1148 | |||
M14963.1 | - | 8 | 1138 | |||
M23204.1 | - | 9 | 1145 | |||
Y07511.1 | - | 8 | 1134 |