Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133995.4 | NP_598756.1 | 2 | 273 | ||
GenBank mRNA | AK149373.1 | - | 2 | 170 | ||
AK149480.1 | - | 2 | 170 | |||
AK165429.1 | - | 2 | 204 | |||
BC021388.1 | - | 2 | 158 | 5055165 |