Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175329.3 | NP_780538.2 | 3 | 692 | ||
GenBank mRNA | AF222853.1 | - | 2 | 208 | ||
AK075746.1 | - | 3 | 373 | |||
AK131581.1 | - | 3 | 391 | |||
AY238604.1 | - | 3 | 692 | |||
BC061190.1 | - | 3 | 399 | 6595773 | ||
BC083357.1 | - | 4 | 882 |