Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159591.1 | NP_001153063.1 | 2, 3 | 511 | ||
NM_134002.2 | NP_598763.1 | 2, 3 | 607 | |||
GenBank mRNA | AK077076.1 | - | 2, 3 | 491 | ||
AK132871.1 | - | 2, 3 | 512 | |||
AK152413.1 | - | 2, 3 | 607 | |||
AK194724.1 | - | 1, 2 | 323 | |||
AK204843.1 | - | 1, 2 | 105 | |||
AK207718.1 | - | 1, 2 | 105 | |||
AK208779.1 | - | 1, 2 | 105 | |||
AK213839.1 | - | 1, 2 | 106 | |||
BC004839.1 | - | 2, 3 | 549 | 2648652 | ||
BC005750.1 | - | 1, 2 | 215 | 3600496 | ||
BC035440.1 | - | 2, 3 | 494 |