Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301302.1 | NP_001288231.1 | 3 | 469 | ||
NM_002537.3 | NP_002528.1 | 3 | 472 | |||
GenBank mRNA | AF057297.1 | - | 3 | 390 | ||
AF242520.1 | - | 3 | 367 | |||
AF242521.1 | - | 3 | 366 | |||
BI597670.1 | - | 3 | 407 | |||
BX648172.1 | - | 2 | 5629 | |||
DA210449.1 | - | 3 | 472 |