Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134010.2 | NP_598771.1 | 24 | 2261 | ||
GenBank mRNA | AK088069.1 | - | 24 | 2262 | ||
AK088303.1 | - | 24 | 2238 | |||
AK088625.1 | - | 24 | 2231 | |||
AK133938.1 | - | 24 | 2240 | |||
AK161665.1 | - | 24 | 2249 | |||
BC004655.1 | - | 20 | 1736 | |||
BC057591.1 | - | 24 | 2228 | 6402677 | ||
BC092261.1 | - | 24 | 2238 | 2646396 |