Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134012.3 | NP_598773.2 | 13, 14 | 1716 | ||
GenBank mRNA | AK028503.1 | - | 6, 7 | 658 | ||
AK033238.1 | - | 8, 9 | 4589 | |||
AK161836.1 | - | 1, 2 | 99 | |||
BC020018.1 | - | 9, 10 | 1027 | 2646754 | ||
BC062907.1 | - | 13, 14 | 1716 | 6813849 | ||
BC064014.1 | - | 15, 16 | 1820 | 6335954 |