Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134012.3 | NP_598773.2 | 7 | 915 | ||
GenBank mRNA | AK033238.1 | - | 2 | 3788 | ||
AK141155.1 | - | 7 | 910 | |||
AK153919.1 | - | 9 | 750 | |||
BC020018.1 | - | 3 | 226 | 2646754 | ||
BC062907.1 | - | 7 | 915 | 6813849 | ||
BC064014.1 | - | 9 | 1019 | 6335954 |