Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134013.3 | NP_598774.2 | 31, 32 | 3806 | ||
GenBank mRNA | AF296169.1 | - | 31, 32 | 3517 | ||
AK140717.1 | - | 11, 12 | 3971 | |||
BC031174.1 | - | 3, 4 | 250 | |||
BC071256.1 | - | 2, 3 | 139 | |||
BC141381.1 | - | 31, 32 | 3519 | |||
CX566261.1 | - | 3, 4 | 194 |