Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035226.1 | NP_001030303.1 | 15 | 1609 | ||
NM_134014.3 | NP_598775.2 | 15 | 1847 | |||
GenBank mRNA | AK136538.1 | - | 8 | 1010 | ||
BC012276.1 | - | 2 | 71 | 3156478 | ||
BC025628.1 | - | 1 | 31 | 5321933 | ||
BC029714.1 | - | 13 | 1370 | |||
BC034710.1 | - | 14 | 1603 | |||
BC062912.1 | - | 15 | 1609 | 6821729 | ||
CF538023.1 | - | 4 | 450 |