Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134023.1 | NP_598784.1 | 9 | 1491 | ||
GenBank mRNA | AK140872.1 | - | 9 | 1503 | ||
AK153677.1 | - | 1279 | ||||
AK220243.1 | - | 2039 | ||||
BC018300.1 | - | 9 | 1491 | 4193046 | ||
BC029773.1 | - | 7 | 1055 | |||
BC037230.1 | - | 9 | 1474 | 2650923 |