Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuedc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172099.1 | NP_001165570.1 | 5 | 1081 | ||
NM_198013.3 | NP_932130.2 | 6 | 1279 | |||
GenBank mRNA | AK049250.1 | - | 6 | 1243 | ||
AK162604.1 | - | 5 | 661 | |||
BC024476.1 | - | 6 | 1128 | 4976954 | ||
BC024529.1 | - | 2 | 80 | |||
BC025434.1 | - | 6 | 1133 | |||
BC029708.1 | - | 6 | 1212 | 5041450 | ||
BC131989.1 | - | 5 | 1052 | 40130266 | ||
BC131991.1 | - | 5 | 1052 | 40130268 | ||
BC145206.1 | - | 5 | 1052 | 9053757 | ||
CF950088.1 | - | 2 | 141 | |||
CK332869.1 | - | 5 | 525 |