Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134029.2 | NP_598790.1 | 4 | 547 | ||
GenBank mRNA | AK008226.1 | - | 515 | |||
AK035283.1 | - | 546 | ||||
AK186031.1 | - | 271 | ||||
AY061970.1 | - | 4 | 534 | |||
BC020084.1 | - | 4 | 547 | 3986525 |