Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134032.2 | NP_598793.2 | 1 | 285 | ||
GenBank mRNA | BC138494.1 | - | 1 | 204 | 8861514 | |
BC138495.1 | - | 1 | 204 | 8861515 |