Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007596.2 | NP_001007597.1 | 3 | 517 | ||
NM_001286448.1 | NP_001273377.1 | 3 | 1062 | |||
NM_153457.7 | NP_703187.2 | 5 | 2598 | |||
GenBank mRNA | AB074899.1 | - | 5 | 2095 | ||
AK002997.1 | - | 3 | 515 | |||
AK013731.1 | - | 3 | 452 | |||
AK014358.1 | - | 3 | 470 | |||
AK132364.1 | - | 5 | 2598 | |||
BC027091.1 | - | 3 | 397 | |||
BC030455.1 | - | 5 | 2150 | 5401661 | ||
BC053926.1 | - | 5 | 2226 | 6312477 | ||
BC058579.1 | - | 5 | 2260 | 6490092 | ||
CT010392.1 | - | 5 | 2095 |