Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080435.1 | NP_536683.1 | 2 | 157 | ||
GenBank mRNA | AF442771.1 | - | 2 | 157 | ||
AK141767.1 | - | 2 | 228 | |||
AK161880.1 | - | 2 | 176 | |||
AK220443.1 | - | 1 | 53 | |||
BC015299.1 | - | 2 | 157 | 4236007 |