Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023794.2 | NP_076283.2 | 7 | 690 | ||
GenBank mRNA | AK018368.1 | - | 7 | 742 | ||
AK031452.1 | - | 7 | 737 | |||
AK049234.1 | - | 7 | 740 | |||
AK080674.1 | - | 7 | 667 | |||
AK164091.1 | - | 7 | 740 | |||
AK168881.1 | - | 7 | 738 | |||
AY004174.1 | - | 6 | 567 | |||
BC034680.1 | - | 7 | 690 | 4036564 |