Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172549.3 | NP_766137.2 | 5 | 713 | ||
GenBank mRNA | AK168811.1 | - | 5 | 673 | ||
BC039933.1 | - | 5 | 540 | |||
BC057551.1 | - | 5 | 641 | |||
BC150825.1 | - | 4 | 277 | 9087736 |