Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172549.3 | NP_766137.2 | 17 | 2856 | ||
GenBank mRNA | BC057551.1 | - | 17 | 2784 | ||
BC150825.1 | - | 16 | 2420 | 9087736 |