Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038695.3 | NP_001033784.2 | 26 | 4041 | ||
NM_173001.3 | NP_766589.1 | 25 | 4068 | |||
GenBank mRNA | AK007123.1 | - | 2 | 173 | ||
AK129204.1 | - | 26 | 4029 | |||
BC005725.1 | - | 5 | 586 | |||
BC026605.1 | - | 11 | 1656 | |||
BC031158.1 | - | 14 | 2128 | |||
BC031200.1 | - | 14 | 2112 | |||
BC031909.1 | - | 9 | 1196 | |||
BC049876.1 | - | 19 | 3111 | |||
BC059264.1 | - | 25 | 3951 | 6409698 | ||
DQ323991.1 | - | 22 | 3506 | |||
KJ461739.1 | - | 16 | 2372 |