Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027874.2 | NP_082150.1 | 8 | 1421 | ||
NM_139059.2 | NP_620690.1 | 8 | 1421 | |||
GenBank mRNA | AK004606.1 | - | 8 | 1181 | ||
AK051052.1 | - | 8 | 1420 | |||
AK088642.1 | - | 8 | 1156 | |||
AK140317.1 | - | 8 | 1146 | |||
AK143534.1 | - | 8 | 1122 | |||
AK146600.1 | - | 8 | 1411 | |||
AK150387.1 | - | 8 | 1150 | |||
AK150815.1 | - | 8 | 1148 | |||
AK152721.1 | - | 8 | 1363 | |||
AK157812.1 | - | 8 | 1156 | |||
AK193235.1 | - | 2 | 106 | |||
BC004604.1 | - | 8 | 1407 | 3585286 |