Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ODF2 SIRNA Assays ›
Gene Symbol:
ODF2
Gene Name:
outer dense fiber of sperm tails 2
Gene Aliases:
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location:
Chr.9: 128455155 - 128501292 on Build GRCh38
UniGene ID:
Hs.129055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9828
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ODF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242352.1
NP_001229281.1
13, 14
1783
NM_001242353.1
NP_001229282.1
13, 14
1869
NM_001242354.1
NP_001229283.1
12, 13
1555
NM_002540.4
NP_002531.3
15, 16
2071
NM_153432.1
NP_702910.1
12, 13
1629
NM_153433.1
NP_702911.1
13, 14
1796
NM_153435.1
NP_702913.1
13, 14
1689
NM_153436.1
NP_702914.1
12, 13
1453
NM_153437.2
NP_702915.1
13, 14
1719
NM_153439.1
NP_702917.1
12, 13
1572
NM_153440.1
NP_702918.1
12, 13
1499
XM_005252010.2
XP_005252067.1
12, 13
1581
XM_005252011.2
XP_005252068.1
12, 13
1521
XM_005252012.3
XP_005252069.1
13, 14
1771
XM_005252013.3
XP_005252070.1
13, 14
1718
XM_005252014.3
XP_005252071.1
13, 14
1767
XM_005252017.4
XP_005252074.1
13, 14
1672
XM_005252018.2
XP_005252075.1
14, 15
1877
XM_005252019.3
XP_005252076.1
14, 15
1896
XM_005252022.2
XP_005252079.1
13, 14
1695
XM_005252023.2
XP_005252080.1
13, 14
1666
XM_005252024.3
XP_005252081.1
13, 14
1685
XM_005252026.4
XP_005252083.1
13, 14
1614
XM_005252029.2
XP_005252086.1
13, 14
1695
XM_006717126.3
XP_006717189.1
13, 14
1771
XM_011518722.2
XP_011517024.1
13, 14
1695
XM_011518723.2
XP_011517025.1
13, 14
1638
XM_011518724.2
XP_011517026.1
14, 15
1859
XM_011518725.2
XP_011517027.1
14, 15
1863
XM_011518726.2
XP_011517028.1
14, 15
1881
XM_011518728.2
XP_011517030.1
13, 14
1632
XM_011518729.2
XP_011517031.1
13, 14
1568
XM_011518730.2
XP_011517032.1
13, 14
1695
XM_011518731.2
XP_011517033.1
12, 13
1524
XM_011518732.2
XP_011517034.1
13, 14
1481
XM_011518733.2
XP_011517035.1
12, 13
1467
XM_011518744.1
XP_011517046.1
12, 13
1521
XM_017014764.1
XP_016870253.1
14, 15
1852
XM_017014765.1
XP_016870254.1
14, 15
1871
XM_017014766.1
XP_016870255.1
12, 13
1581
XM_017014767.1
XP_016870256.1
14, 15
1863
XM_017014768.1
XP_016870257.1
13, 14
1655
XM_017014769.1
XP_016870258.1
13, 14
1684
XM_017014770.1
XP_016870259.1
14, 15
1839
XM_017014771.1
XP_016870260.1
14, 15
1820
XM_017014772.1
XP_016870261.1
14, 15
1896
XM_017014773.1
XP_016870262.1
14, 15
1872
XM_017014774.1
XP_016870263.1
13, 14
1582
XM_017014775.1
XP_016870264.1
14, 15
1839
XM_017014776.1
XP_016870265.1
14, 15
1820
XM_017014777.1
XP_016870266.1
13, 14
1622
XM_017014778.1
XP_016870267.1
13, 14
1725
XM_017014779.1
XP_016870268.1
12, 13
1466
XM_017014780.1
XP_016870269.1
13, 14
1649
XM_017014781.1
XP_016870270.1
12, 13
1404
GenBank mRNA
AF012549.1
-
13, 14
1659
AF053970.1
-
11, 12
1358
AK093367.1
-
12, 13
1579
AK299303.1
-
13, 14
1631
AK300793.1
-
12, 13
1404
AK301754.1
-
12, 13
1496
AK301842.1
-
12, 13
1421
AK301855.1
-
12, 13
1475
AK302684.1
-
13, 14
1723
AY319414.1
-
12, 13
1605
AY366499.1
-
13, 14
1566
BC010629.1
-
15, 16
1869
3874501
BC091500.1
-
13, 14
1683
30378678
DQ444713.1
-
11, 12
1396
DQ444714.1
-
11, 12
1339
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
multicellular organism development
spermatid development
structural molecule activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-gq622:80/100.66.78.111:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline