Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023894.1 | NP_076383.1 | 3, 4 | 682 | ||
GenBank mRNA | AF201698.1 | - | 3, 4 | 682 | ||
AK018818.1 | - | 2, 3 | 123 | |||
BC099451.1 | - | 3, 4 | 621 | 30716051 | ||
DQ058646.1 | - | 3, 4 | 682 |