Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027088.2 | NP_081364.1 | 9 | 810 | ||
GenBank mRNA | AB047820.1 | - | 9 | 711 | ||
AK079374.1 | - | 9 | 826 | |||
AK129108.1 | - | 9 | 718 | |||
AK170576.1 | - | 9 | 811 | |||
AK171561.1 | - | 9 | 812 | |||
AK185783.1 | - | 3 | 195 | |||
AK189418.1 | - | 3 | 196 | |||
AK193408.1 | - | 3 | 195 | |||
BC042489.1 | - | 9 | 761 | |||
BC050901.1 | - | 9 | 773 | 6402443 |