Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024233.3 | NP_077195.2 | 5 | 547 | ||
GenBank mRNA | AK007731.1 | - | 5 | 549 | ||
AK150287.1 | - | 5 | 463 | |||
AK171619.1 | - | 5 | 450 | |||
AK172641.1 | - | 5 | 525 | |||
AK182577.1 | - | 2 | 89 | |||
AK185600.1 | - | 2 | 89 | |||
BC003445.1 | - | 5 | 483 | 2651110 |