Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 0610010K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177601.1 | NP_001171072.1 | 4 | 501 | ||
NM_001177603.1 | NP_001171074.1 | 4 | 501 | |||
NM_001177606.1 | NP_001171077.1 | 3 | 399 | |||
NM_001177607.1 | NP_001171078.1 | 3 | 399 | |||
NM_026757.2 | NP_081033.1 | 3 | 399 | |||
NM_145758.1 | NP_665701.1 | 4 | 501 | |||
GenBank mRNA | AK003842.1 | - | 3 | 399 | ||
AK169173.1 | - | 3 | 390 | |||
AK170380.1 | - | 4 | 499 | |||
AK178853.1 | - | 219 | ||||
AK208696.1 | - | 229 | ||||
BC055357.1 | - | 4 | 441 | 5325264 | ||
BE861439.1 | - | 3 | 338 | |||
CB192659.1 | - | 4 | 502 | |||
CK791552.1 | - | 4 | 480 | |||
EL959640.1 | - | 4 | 446 |