Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134034.2 | NP_598795.2 | 12 | 2187 | ||
GenBank mRNA | AK037253.1 | - | 12 | 2119 | ||
AK041513.1 | - | 12 | 2132 | |||
AK044742.1 | - | 12 | 2130 | |||
AK090014.1 | - | 12 | 2163 | |||
AK129344.1 | - | 12 | 2098 | |||
AK132660.1 | - | 12 | 1709 | |||
AK133211.1 | - | 3 | 303 | |||
AK165940.1 | - | 11 | 2070 | |||
AK166627.1 | - | 12 | 2112 | |||
BC006870.1 | - | 12 | 2098 | 3595439 |