Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipas39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142580.1 | NP_001136052.1 | 13 | 1000 | ||
NM_001142581.1 | NP_001136053.1 | 12 | 943 | |||
NM_134044.3 | NP_598805.2 | 12 | 957 | |||
GenBank mRNA | AK046322.1 | - | 13 | 971 | ||
AK049643.1 | - | 13 | 947 | |||
AK079055.1 | - | 13 | 947 | |||
AK082261.1 | - | 13 | 928 | |||
AK145043.1 | - | 983 | ||||
BC016646.1 | - | 12 | 866 | 2651233 | ||
BC023716.1 | - | 12 | 968 | 5346489 |