Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178912.3 | NP_849243.2 | 4 | 870 | ||
GenBank mRNA | AK039944.1 | - | 4 | 874 | ||
AK050306.1 | - | 4 | 871 | |||
AK053715.1 | - | 4 | 870 | |||
AK084697.1 | - | 4 | 874 | |||
AK086395.1 | - | 4 | 873 | |||
AK173211.1 | - | 4 | 849 | |||
BC150785.1 | - | 4 | 864 | 9087696 |