Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289572.1 | NP_001276501.1 | 3 | 289 | ||
NM_001289573.1 | NP_001276502.1 | 4 | 385 | |||
NM_001289574.1 | NP_001276503.1 | 4 | 451 | |||
NM_178914.4 | NP_849245.2 | 4 | 385 | |||
GenBank mRNA | AK036359.1 | - | 4 | 369 | ||
AK077500.1 | - | 4 | 296 | |||
AK153698.1 | - | 4 | 361 | |||
AK161308.1 | - | 3 | 235 | |||
AK169473.1 | - | 4 | 366 | |||
BC046960.1 | - | 4 | 343 | 6391450 | ||
BY252962.1 | - | 4 | 384 | |||
DQ196101.1 | - | 3 | 115 |