Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014057.4 | NP_054776.1 | 5 | 727 | ||
NM_024416.4 | NP_077727.3 | 5 | 646 | |||
NM_033014.3 | NP_148935.1 | 5 | 1009 | |||
GenBank mRNA | AF086912.1 | - | 5 | 574 | ||
AF088023.1 | - | 2 | 65 | |||
AF100758.1 | - | 5 | 574 | |||
AF202167.1 | - | 4 | 499 | |||
AK075521.1 | - | 4 | 538 | |||
AK129845.1 | - | 4 | 531 | |||
AK289628.1 | - | 5 | 724 | |||
AK295433.1 | - | 5 | 646 | |||
AL110267.1 | - | 4 | 516 | |||
BC037273.2 | - | 4 | 515 | 5265880 | ||
BC095443.1 | - | 4 | 531 | 30405707 | ||
BG772245.1 | - | 5 | 650 | |||
BX646984.1 | - | 5 | 684 | |||
CR533516.1 | - | 4 | 448 |