Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014057.4 | NP_054776.1 | 2 | 371 | ||
NM_024416.4 | NP_077727.3 | 2 | 290 | |||
NM_033014.3 | NP_148935.1 | 2 | 653 | |||
GenBank mRNA | AF086912.1 | - | 2 | 218 | ||
AF100758.1 | - | 2 | 218 | |||
AF202167.1 | - | 1 | 143 | |||
AK075521.1 | - | 1 | 182 | |||
AK129845.1 | - | 1 | 175 | |||
AK289628.1 | - | 2 | 368 | |||
AK295433.1 | - | 2 | 290 | |||
AL110267.1 | - | 1 | 160 | |||
BC037273.2 | - | 1 | 159 | 5265880 | ||
BC095443.1 | - | 1 | 175 | 30405707 | ||
BG772245.1 | - | 2 | 292 | |||
BX646984.1 | - | 2 | 328 | |||
CR533516.1 | - | 1 | 92 |