Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023697.2 | NP_076186.1 | 2 | 749 | ||
GenBank mRNA | AF303831.1 | - | 2 | 764 | ||
AK142290.1 | - | 2 | 448 | |||
BC020094.1 | - | 2 | 737 | 4014290 | ||
BC092299.1 | - | 2 | 745 | 5060293 |