Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACVR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106.3 | NP_001097.2 | 2 | 238 | ||
XM_005265583.3 | XP_005265640.1 | 2 | 2700 | |||
XM_017007514.1 | XP_016863003.1 | 2 | 2679 | |||
XM_017007515.1 | XP_016863004.1 | 2 | 324 | |||
XM_017007516.1 | XP_016863005.1 | 2 | 3648 | |||
GenBank mRNA | AB209449.1 | - | 1 | 4003 | ||
BC096243.1 | - | 2 | 218 | 40005760 | ||
BC096244.1 | - | 2 | 218 | 40005761 | ||
BC096245.1 | - | 2 | 218 | 40005763 | ||
BC099642.4 | - | 2 | 218 | 40005762 | ||
BP352716.1 | - | 2 | 201 | |||
X77533.1 | - | 2 | 218 |