Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134065.4 | NP_598826.3 | 1, 2 | 539 | ||
GenBank mRNA | AF353717.1 | - | 1, 2 | 352 | ||
AK034083.1 | - | 1, 2 | 508 | |||
AK038733.1 | - | 1, 2 | 514 | |||
AK046185.1 | - | 1, 2 | 514 | |||
AK160413.1 | - | 1, 2 | 474 | |||
AK166252.1 | - | 1, 2 | 480 | |||
BC014708.1 | - | 1, 2 | 494 | 4223134 |