Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam149b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024512.2 | NP_001019683.1 | 3 | 366 | ||
NM_001177633.1 | NP_001171104.1 | 3 | 366 | |||
NM_172379.3 | NP_759011.2 | 3 | 366 | |||
GenBank mRNA | AK031973.1 | - | 3 | 369 | ||
AK037808.1 | - | 3 | 254 | |||
AK077495.1 | - | 3 | 269 | |||
BB622169.1 | - | 3 | 368 | |||
BC025183.1 | - | 3 | 262 | 5011464 | ||
BC057907.1 | - | 3 | 268 | 5064176 | ||
BC058650.1 | - | 3 | 209 | |||
BC059221.1 | - | 3 | 209 | |||
BC069846.1 | - | 3 | 264 | 4196478 | ||
BC080849.1 | - | 3 | 306 | 5717789 |