Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177992.2 | NP_818773.1 | 3 | 268 | ||
GenBank mRNA | AK017821.1 | - | 3 | 563 | ||
AK031004.1 | - | 3 | 270 | |||
AK075772.1 | - | 3 | 226 | |||
AK163048.1 | - | 1 | 717 | |||
BC003886.1 | - | 3 | 227 | 3590411 | ||
BC024109.1 | - | 3 | 224 | 5149353 | ||
BY099107.1 | - | 3 | 269 |