Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153127.3 | NP_694767.3 | 6 | 1521 | ||
GenBank mRNA | AK077884.1 | - | 6 | 1523 | ||
AK078470.1 | - | 6 | 1497 | |||
AK134285.1 | - | 6 | 1560 | |||
BC145845.1 | - | 6 | 1441 | 40131148 |