Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175341.4 | NP_780550.1 | 5 | 1331 | ||
NM_207515.2 | NP_997398.1 | 5 | 1331 | |||
GenBank mRNA | AK028797.1 | - | 5 | 1296 | ||
AK085709.1 | - | 5 | 1294 | |||
AK151913.1 | - | 5 | 1295 | |||
AK151926.1 | - | 5 | 1295 | |||
AK151957.1 | - | 5 | 1295 | |||
AK153261.1 | - | 5 | 1185 | |||
AK159213.1 | - | 5 | 1289 | |||
AK164372.1 | - | 5 | 1294 | |||
AK167479.1 | - | 5 | 1161 | |||
AK167694.1 | - | 6 | 1096 | |||
AK220470.1 | - | 1305 | ||||
BC020406.1 | - | 4 | 729 | |||
BC023895.1 | - | 5 | 1283 | |||
BC026711.1 | - | 4 | 696 | |||
BC075665.1 | - | 4 | 934 | 30605146 | ||
BC079898.1 | - | 5 | 1280 | |||
GQ906534.1 | - | 4 | 649 |