Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047433.2 | NP_001040898.1 | 2 | 532 | ||
NM_172254.4 | NP_758458.1 | 3 | 607 | |||
GenBank mRNA | AK011448.1 | - | 3 | 525 | ||
AK017764.1 | - | 3 | 650 | |||
AK048097.1 | - | 3 | 497 | |||
BC029910.1 | - | 3 | 498 | 4920779 |