Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thtpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153083.5 | NP_694723.1 | 2 | 1262 | ||
GenBank mRNA | AF432864.2 | - | 2 | 1093 | ||
AK085173.1 | - | 2 | 958 | |||
AK132479.1 | - | 2 | 954 | |||
BC025562.1 | - | 2 | 1246 | 5322971 |