Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170694.1 | NP_001164165.1 | 13 | 1566 | ||
NM_134083.4 | NP_598844.3 | 13 | 1561 | |||
GenBank mRNA | AK013540.1 | - | 1671 | |||
AK031242.1 | - | 13 | 1527 | |||
AK031248.1 | - | 13 | 1532 | |||
AK147784.1 | - | 14 | 1567 | |||
AK160212.1 | - | 2 | 105 | |||
AK160881.1 | - | 14 | 1753 | |||
AK183696.1 | - | 2 | 130 | |||
AK191301.1 | - | 2 | 118 | |||
AK204057.1 | - | 3 | 196 | |||
AK210358.1 | - | 3 | 196 | |||
AK217036.1 | - | 3 | 196 | |||
BC003224.1 | - | 11 | 1359 | 3482729 | ||
JN086338.1 | - | 12 | 1482 | |||
JN086339.1 | - | 13 | 1720 |