Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166456.1 | NP_001159928.1 | 14, 15 | 1574 | ||
NM_001166458.1 | NP_001159930.1 | 14, 15 | 1781 | |||
NM_134086.4 | NP_598847.2 | 14, 15 | 2267 | |||
GenBank mRNA | AF184240.2 | - | 13, 14 | 1359 | ||
AK034130.1 | - | 14, 15 | 2269 | |||
AK035098.1 | - | 13, 14 | 1286 | |||
AK047459.1 | - | 14, 15 | 1798 | |||
AK049173.1 | - | 14, 15 | 1483 | |||
AK049294.1 | - | 14, 15 | 1547 | |||
AK081724.1 | - | 14, 15 | 1783 | |||
AK082683.1 | - | 14, 15 | 1485 | |||
AK162612.1 | - | 14, 15 | 1632 | |||
AK163914.1 | - | 9, 10 | 885 | |||
AK165188.1 | - | 14, 15 | 1834 | |||
BC030378.1 | - | 14, 15 | 1483 | 5375319 |