Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134089.1 | NP_598850.1 | 15 | 2085 | ||
GenBank mRNA | AF441233.1 | - | 15 | 2085 | ||
AK122211.1 | - | 15 | 2139 | |||
BC060689.1 | - | 15 | 2075 | 6417366 | ||
BC062888.1 | - | 15 | 2077 | 5721250 |