Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134090.2 | NP_598851.2 | 4 | 735 | ||
GenBank mRNA | AK136991.1 | - | 4 | 719 | ||
BC011472.1 | - | 4 | 675 | 4168514 | ||
BC024420.1 | - | 4 | 668 | 4985334 |